Assay Details
Target Gene Details
Entrez Gene ID: | 12048 |
Gene Name: | BCL2-like 1 |
Gene Aliases: |
Bcl(X)L, Bcl-XL, Bcl2l, BclX, bcl-x, bcl2-L-1 |
Location: |
Chr.2:152758732-152832581 on Build GRCm38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bcl2l1 | NM_001289739.1 | NP_001276668.1 | ||
| U51277.1 | AAC53458.1 |
Target Gene Details
Entrez Gene ID: | 84682 |
Gene Name: | cytochrome c oxidase subunit IV isoform 2 |
Gene Aliases: |
Cox4b, CoxIV-2 |
Location: |
Chr.2:152754173-152765037 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cox4i2 | NM_053091.2 | NP_444321.1 | ||
| AF271382.1 | AAK49433.1 | |||
| BC049623.1 | AAH49623.1 |
More Information
Set Membership: |
|

Genomic Map