Assay Details
Target Gene Details
Entrez Gene ID: | 12831 |
Gene Name: | collagen, type V, alpha 1 |
Gene Aliases: |
AI413331 |
Location: |
Chr.2:27886395-28039510 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Col5a1 | NM_015734.2 | NP_056549.2 | ||
| AB009993.1 | BAA28786.1 | |||
| BC138447.1 | ||||
| BC138449.1 | ||||
| S63661.1 |
More Information
Set Membership: |
|

Genomic Map