Assay Details
Target Gene Details
Entrez Gene ID: | 77771 |
Gene Name: | cysteine-serine-rich nuclear protein 3 |
Gene Aliases: |
A330102K23Rik, CSRNP-3, Csnrp3, Mbu1, taip-2 |
Location: |
Chr.2:65845767-66031546 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Csrnp3 | NM_001290666.1 | NP_001277595.1 | ||
| NM_153409.5 | NP_700458.3 | |||
| NM_178634.2 | NP_848749.2 | |||
| AK020730.1 | 3 | 322 | ||
| AK039150.1 | BAC30257.1 | |||
| AK080761.1 | 3 | 322 | ||
| AK082649.1 | BAC38559.1 | |||
| BC055701.1 | AAH55701.1 | |||
| BY297754.1 | ||||
| EF210820.1 |
More Information
Set Membership: |
|

Genomic Map