Assay Details
Target Gene Details
Entrez Gene ID: | 13012 |
Gene Name: | cystatin 8 (cystatin-related epididymal spermatogenic) |
Gene Aliases: |
Cres, Cst-rs1 |
Location: |
Chr.2:148798839-148805596 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cst8 | NM_009978.2 | 4 | 581 | NP_034108.1 |
| AF090691.1 | 4 | 570 | AAC36316.1 | |
| AK018943.1 | 4 | 582 | BAB31489.1 | |
| AK054409.1 | 4 | 564 | BAC35768.1 | |
| AK161808.1 | 4 | 574 | BAE36585.1 | |
| BC049753.1 | 4 | 586 | AAH49753.1 | |
| S49926.1 | 4 | 382 | AAC35390.1 |

Genomic Map