Assay Details
Target Gene Details
Entrez Gene ID: | 228889 |
Gene Name: | DEAD (Asp-Glu-Ala-Asp) box polypeptide 27 |
Gene Aliases: |
C86129 |
Location: |
Chr.2:167015255-167034950 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ddx27 | NM_153065.3 | 11 | 1211 | NP_694705.2 |
| AK134274.1 | 11 | 1212 | BAE22077.1 | |
| AK138442.1 | 11 | 1235 | BAE23664.1 | |
| AK146469.1 | 11 | 1239 | ||
| BC011321.1 | 10 | 1088 | AAH11321.1 |

Genomic Map