Assay Details
Target Gene Details
Entrez Gene ID: | 227525 |
Gene Name: | DNA cross-link repair 1C, PSO2 homolog (S. cerevisiae) |
Gene Aliases: |
9930121L06Rik, AI661365, Art, Snm1l |
Location: |
Chr.2:3424062-3474330 on Build GRCm38 |
Assay Gene Location: | Within Intron 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dclre1c | NM_001110214.1 | NP_001103684.1 | ||
| NM_175683.4 | NP_783614.1 | |||
| AK037126.1 | BAC29713.1 | |||
| AK052369.1 | BAC34960.1 | |||
| AK088810.1 | BAC40586.1 | |||
| BC108935.2 | AAI08936.1 |
Target Gene Details
Entrez Gene ID: | 64707 |
Gene Name: | suppressor of variegation 3-9 homolog 2 (Drosophila) |
Gene Aliases: |
4930507K23Rik, AA536750, D030054H19Rik, D2Ertd544e, KMT1B |
Location: |
Chr.2:3455815-3474986 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Suv39h2 | NM_022724.4 | 6 | 2618 | NP_073561.2 |
| NR_027509.1 | 5 | 2472 | ||
| AK083457.1 | 6 | 2596 | BAC38921.1 |
More Information
Set Membership: |
|

Genomic Map