Assay Details
Target Gene Details
Entrez Gene ID: | 78655 |
Gene Name: | eukaryotic translation initiation factor 3, subunit J1 |
Gene Aliases: |
1810016I04Rik, 2700079K05Rik, AA409117, AA409446, C78575, Eif3j, Eif3j-1, Eif3j-2, Eif3j2, Eif3s1, eIF-3-alpha-A, eIF-3-alpha-B, eIF3-alpha, eIF3-p35, eIF3j-A, eIF3j-B |
Location: |
Chr.2:122028113-122053632 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Eif3j1 | NM_144545.4 | NP_653128.2 | ||
| AK082048.1 | BAC38399.1 | |||
| AK147154.1 | BAE27721.1 | |||
| AK148004.1 | BAE28282.1 | |||
| AK154934.1 | BAE32935.1 | |||
| AK161038.1 | BAE36162.1 | |||
| BC006055.1 | AAH06055.1 |
More Information
Set Membership: |
|

Genomic Map