Assay Details
Target Gene Details
Entrez Gene ID: | 98221 |
Gene Name: | eukaryotic translation initiation factor 3, subunit M |
Gene Aliases: |
Ga17, Pcid1, Tango7 |
Location: |
Chr.2:104999656-105017027 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Eif3m | NM_145380.2 | NP_663355.1 | ||
| AK077480.1 | BAC36821.1 | |||
| AK084738.1 | BAC39270.1 | |||
| AK146222.1 | BAE26990.1 | |||
| AK168066.1 | BAE40042.1 | |||
| BC005598.1 | AAH05598.1 | |||
| BC089568.1 | AAH89568.1 | |||
| BC103795.1 | AAI03796.1 | |||
| BC116787.1 | ||||
| BC116789.1 |

Genomic Map