Assay Details
Target Gene Details
Entrez Gene ID: | 14134 |
Gene Name: | ficolin B |
Gene Aliases: |
Fcn2 |
Location: |
Chr.2:28076479-28084913 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fcnb | NM_010190.1 | NP_034320.1 | ||
| AF063217.1 | AAC98781.1 | |||
| AK010913.1 | ||||
| AK151676.1 | BAE30601.1 | |||
| BC107220.1 | AAI07221.1 | |||
| BC107221.1 | AAI07222.1 |
More Information
Set Membership: |
|

Genomic Map