Assay Details
Target Gene Details
| Entrez Gene ID: | 71436 | 
| Gene Name: | fibronectin leucine rich transmembrane protein 3 | 
| Gene Aliases: | 5530600M07Rik, C430047I10Rik, mKIAA1469 | 
| Location: | Chr.2:140658198-140671476 on Build GRCm38 | 
| Assay Gene Location: | Within Exon 2 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| Flrt3 | NM_001172160.1 | 2 | 329 | NP_001165631.1 | 
| NM_178382.4 | 2 | 329 | NP_848469.1 | |
| AK028252.1 | 2 | 323 | BAC25843.1 | |
| AK031464.1 | 2 | 323 | BAC27417.1 | |
| AK129369.1 | 2 | 253 | BAC98179.1 | |
| AK161100.1 | 2 | 323 | ||
| BC052043.1 | 2 | 304 | AAH52043.1 | 
Target Gene Details
| Entrez Gene ID: | 72899 | 
| Gene Name: | MACRO domain containing 2 | 
| Gene Aliases: | 1110033L15Rik, 2610107G07Rik, 2900006F19Rik | 
| Location: | Chr.2:140395356-142392966 on Build GRCm38 | 
| Assay Gene Location: | Within Intron 5 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| Macrod2 | NM_001013802.3 | NP_001013824.2 | ||
| AK013491.1 | ||||
| AK134694.1 | BAE22244.1 | 
 Back To Top
 Back To TopMore Information
| Set Membership: |  Intragenic  Exonic  Intronic  Non-exonic | 
Gene Ontology Categories:
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top