Assay Details
Target Gene Details
Entrez Gene ID: | 71409 |
Gene Name: | formin-like 2 |
Gene Aliases: |
5430425K04Rik, Man |
Location: |
Chr.2:52857716-53134202 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fmnl2 | NM_172409.2 | NP_765997.2 | ||
| BM941619.1 |

Genomic Map