Assay Details
Target Gene Details
Entrez Gene ID: | 227960 |
Gene Name: | grancalcin |
Gene Aliases: |
5133401E04Rik, AI573844 |
Location: |
Chr.2:62664327-62694109 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gca | NM_145523.3 | NP_663498.1 | ||
| AB088388.1 | BAC07231.1 | |||
| AF518325.1 | AAM66720.1 | |||
| AK033740.1 | BAC28458.1 | |||
| AK046347.1 | BAC32686.1 | |||
| AK049317.1 | BAC33679.1 | |||
| AK050540.1 | BAC34315.1 | |||
| AK078915.1 | BAC37457.1 | |||
| BC021450.1 | AAH21450.1 |

Genomic Map