Assay Details
Target Gene Details
Entrez Gene ID: | 227960 |
Gene Name: | grancalcin |
Gene Aliases: |
5133401E04Rik, AI573844 |
Location: |
Chr.2:62664327-62694109 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gca | NM_145523.3 | 6 | 632 | NP_663498.1 |
| AB088388.1 | 6 | 526 | BAC07231.1 | |
| AF518325.1 | 6 | 616 | AAM66720.1 | |
| AK033740.1 | 6 | 665 | BAC28458.1 | |
| AK046347.1 | 6 | 674 | BAC32686.1 | |
| AK049317.1 | 6 | 633 | BAC33679.1 | |
| AK050540.1 | 6 | 674 | BAC34315.1 | |
| AK078915.1 | 6 | 633 | BAC37457.1 | |
| BC021450.1 | 6 | 626 | AAH21450.1 |

Genomic Map