Assay Details
Target Gene Details
Entrez Gene ID: | 14588 |
Gene Name: | glial cell line derived neurotrophic factor family receptor alpha 4 |
Gene Aliases: |
AW060733, G630015H18Rik |
Location: |
Chr.2:131039632-131043088 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gfra4 | NM_001136063.2 | NP_001129535.1 | ||
| NM_001271001.1 | NP_001257930.1 | |||
| NM_001271002.1 | NP_001257931.1 | |||
| NM_020014.2 | NP_064398.1 | |||
| AB041809.1 | 2 | 963 | BAA95116.1 | |
| AJ276514.1 | CAB89687.1 | |||
| AJ276516.1 | CAB89689.1 | |||
| AJ276870.1 | CAB89690.1 | |||
| AJ276871.1 | CAB89691.1 | |||
| AK090192.1 | BAC41129.1 | |||
| BC111808.1 | AAI11809.1 | |||
| BC141873.1 | ||||
| BC141874.1 |
More Information
Set Membership: |
|

Genomic Map