Assay Details
Target Gene Details
Entrez Gene ID: | 207182 |
Gene Name: | gamma-glutamyltransferase 7 |
Gene Aliases: |
1110017C11Rik, 6330563L03Rik, Ggtl3 |
Location: |
Chr.2:155490379-155514860 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ggt7 | NM_144786.2 | NP_659035.2 | ||
| AK032051.1 | BAC27672.1 | |||
| AK131175.1 | BAD21425.1 | |||
| AK193055.1 | ||||
| BC005772.1 | AAH05772.1 |
More Information
Set Membership: |
|

Genomic Map