Assay Details
Target Gene Details
Entrez Gene ID: | 641368 |
Gene Name: | serine protease inhibitor, Kunitz type 5 |
Gene Aliases: |
Gm14316, OTTMUSG00000016293 |
Location: |
Chr.2:164715305-164718068 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Spint5 | NM_001040055.1 | 3 | 303 | NP_001035144.1 |
| AY542490.1 | 3 | 303 | AAT27478.1 | |
| BC147380.1 | 3 | 285 | ||
| BC147381.1 | 3 | 285 |
More Information
Set Membership: |
|

Genomic Map