Assay Details
Target Gene Details
Entrez Gene ID: | 228366 |
Gene Name: | glycosyltransferase-like 1B |
Gene Aliases: |
5730485C17Rik, AI891893, Large2, Largel, mKIAA4105 |
Location: |
Chr.2:92365046-92371940 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gyltl1b | NM_001166633.2 | NP_001160105.1 | ||
| NM_001290773.1 | NP_001277702.1 | |||
| NM_001290774.1 | NP_001277703.1 | |||
| NM_001290775.1 | NP_001277704.1 | |||
| NM_172670.3 | NP_766258.2 | |||
| AK053214.1 | BAC35314.1 | |||
| AK077630.1 | BAC36913.1 | |||
| AK081797.1 | ||||
| AY742914.1 | AAU95213.1 | |||
| AY742915.1 | AAU95214.1 | |||
| CF170217.1 |
More Information
Set Membership: |
|

Genomic Map