Assay Details
Target Gene Details
Entrez Gene ID: | 15112 |
Gene Name: | hydroxyacid oxidase 1, liver |
Gene Aliases: |
GOX, Gox1, Hao-1 |
Location: |
Chr.2:134497361-134554352 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hao1 | NM_010403.2 | NP_034533.1 | ||
| AF104312.1 | AAD25332.1 | |||
| AK149565.1 | BAE28963.1 | |||
| BC119535.1 | ||||
| BC119536.1 |
More Information
Set Membership: |
|

Genomic Map