Assay Details
Target Gene Details
Entrez Gene ID: | 68526 |
Gene Name: | G protein-coupled receptor 155 |
Gene Aliases: |
1110017O10Rik, DEPDC3, F730029F15Rik, PGR22 |
Location: |
Chr.2:73341506-73386851 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gpr155 | NM_001190297.2 | NP_001177226.1 | ||
| NM_001276443.1 | NP_001263372.1 | |||
| NM_001276444.1 | NP_001263373.1 | |||
| AK089436.1 | ||||
| AK165887.1 | ||||
| BC115698.1 | ||||
| HM161763.2 | ||||
| HM161764.2 | ||||
| HM161765.2 | ||||
| HM161766.2 | ||||
| KF358283.1 | ||||
| KF358284.1 |
More Information
Set Membership: |
|

Genomic Map