Assay Details
Target Gene Details
Entrez Gene ID: | 50915 |
Gene Name: | growth factor receptor bound protein 14 |
Gene Aliases: |
AI505286 |
Location: |
Chr.2:64912476-65022766 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Grb14 | NM_016719.1 | NP_057928.1 | ||
| AF155647.1 | AAF43996.1 | |||
| AK010849.1 | BAB27221.3 | |||
| AK010903.1 | BAB27256.3 | |||
| AK034134.1 | ||||
| AK147041.1 | BAE27629.1 | |||
| BC138581.1 | ||||
| BC145913.1 |
More Information
Set Membership: |
|

Genomic Map