Assay Details
Target Gene Details
Entrez Gene ID: | 227753 |
Gene Name: | gelsolin |
Gene Aliases: |
ADF |
Location: |
Chr.2:35256359-35307902 on Build GRCm38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gsn | NM_001206367.1 | NP_001193296.1 | ||
| NM_001206368.1 | NP_001193297.1 | |||
| NM_001206369.1 | NP_001193298.1 | |||
| NM_146120.4 | NP_666232.2 | |||
| AK076156.1 | BAC36223.1 | |||
| AK089934.1 | BAC41004.1 | |||
| AK135632.1 | ||||
| AK143664.1 | BAE25485.1 | |||
| AK146969.1 | BAE27574.1 | |||
| AK148957.1 | BAE28702.1 | |||
| AK150359.1 | BAE29494.1 | |||
| AK151684.1 | BAE30608.1 | |||
| AK153029.1 | BAE31661.1 | |||
| AK166831.1 | BAE39054.1 | |||
| AK166932.1 | BAE39126.1 | |||
| AK167918.1 | BAE39925.1 | |||
| AK168461.1 | BAE40358.1 | |||
| AK168532.1 | BAE40410.1 | |||
| AK168707.1 | BAE40549.1 | |||
| AK168725.1 | BAE40566.1 | |||
| AK168829.1 | ||||
| AK169041.1 | BAE40830.1 | |||
| AK169670.1 | BAE41291.1 | |||
| AK178123.1 | ||||
| BC023143.1 | ||||
| BC060377.1 | AAH60377.1 | |||
| J04953.1 | AAA37677.1 |
More Information
Set Membership: |
|

Genomic Map