Assay Details
Target Gene Details
Entrez Gene ID: | 227753 |
Gene Name: | gelsolin |
Gene Aliases: |
ADF |
Location: |
Chr.2:35256359-35307902 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gsn | NM_001206367.1 | 10 | 1335 | NP_001193296.1 |
| NM_001206368.1 | 9 | 1251 | NP_001193297.1 | |
| NM_001206369.1 | 9 | 1296 | NP_001193298.1 | |
| NM_146120.4 | 9 | 1358 | NP_666232.2 | |
| AK076156.1 | 9 | 1298 | BAC36223.1 | |
| AK089934.1 | 10 | 1290 | BAC41004.1 | |
| AK135632.1 | 8 | 1071 | ||
| AK143664.1 | 9 | 1325 | BAE25485.1 | |
| AK146969.1 | 10 | 1252 | BAE27574.1 | |
| AK148957.1 | 10 | 1315 | BAE28702.1 | |
| AK150359.1 | 10 | 1258 | BAE29494.1 | |
| AK151684.1 | 10 | 1277 | BAE30608.1 | |
| AK153029.1 | 10 | 1255 | BAE31661.1 | |
| AK166831.1 | 9 | 1271 | BAE39054.1 | |
| AK166932.1 | 10 | 1249 | BAE39126.1 | |
| AK167918.1 | 10 | 1265 | BAE39925.1 | |
| AK168461.1 | 10 | 1268 | BAE40358.1 | |
| AK168532.1 | 10 | 1269 | BAE40410.1 | |
| AK168707.1 | 9 | 1204 | BAE40549.1 | |
| AK168725.1 | 10 | 1252 | BAE40566.1 | |
| AK168829.1 | 9 | 1303 | ||
| AK169041.1 | 10 | 1255 | BAE40830.1 | |
| AK169670.1 | 10 | 1280 | BAE41291.1 | |
| AK200166.1 | ||||
| AK219732.1 | 1 | 33 | ||
| BC023143.1 | 9 | 1296 | ||
| BC060377.1 | 10 | 1283 | AAH60377.1 | |
| J04953.1 | 8 | 1145 | AAA37677.1 |
More Information
Set Membership: |
|

Genomic Map