Assay Details
Target Gene Details
Entrez Gene ID: | 227835 |
Gene Name: | glycosyltransferase-like domain containing 1 |
Gene Aliases: |
E330008O22Rik |
Location: |
Chr.2:44564412-44927918 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gtdc1 | NM_172662.3 | 3 | 316 | NP_766250.2 |
| AK028468.1 | 3 | 239 | BAC25967.1 | |
| AK038057.1 | 2 | 187 | ||
| AK054272.1 | 3 | 317 | BAC35713.1 | |
| AK163943.1 | 3 | 274 | BAE37544.1 |
More Information
Set Membership: |
|

Genomic Map