Assay Details
Target Gene Details
Entrez Gene ID: | 70239 |
Gene Name: | general transcription factor IIIC, polypeptide 5 |
Gene Aliases: |
2700084A09Rik, TFIIIC63, TFiiiC2-63 |
Location: |
Chr.2:28566245-28583746 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gtf3c5 | NM_001290484.1 | 4 | 925 | NP_001277413.1 |
| NM_148928.3 | 4 | 925 | NP_683730.1 | |
| AK045419.1 | 4 | 517 | BAC32354.1 | |
| AK147014.1 | 4 | 903 | BAE27608.1 | |
| BC027055.1 | 4 | 892 | AAH27055.1 | |
| BC027247.1 | 4 | 835 | AAH27247.1 | |
| BC039171.1 | 4 | 846 | AAH39171.1 |
More Information
Set Membership: |
|

Genomic Map