Assay Details
Target Gene Details
Entrez Gene ID: | 140483 |
Gene Name: | histamine N-methyltransferase |
Gene Aliases: |
1500031F01Rik, AI788969 |
Location: |
Chr.2:24002911-24049757 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hnmt | NM_080462.2 | NP_536710.1 | ||
| AB070524.1 | BAB84318.1 | |||
| AF297037.1 | AAL09305.1 | |||
| BC033928.1 | AAH33928.1 |

Genomic Map