Assay Details
Target Gene Details
Entrez Gene ID: | 16396 |
Gene Name: | itchy, E3 ubiquitin protein ligase |
Gene Aliases: |
6720481N21Rik, 8030492O04Rik, A130065M08, AIP4, C230047C07Rik |
Location: |
Chr.2:155133379-155226855 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Itch | NM_001243712.1 | NP_001230641.1 | ||
| NM_008395.3 | NP_032421.2 | |||
| AF037454.1 | AAB99764.1 | |||
| AK037940.1 | ||||
| AK048303.1 | BAC33298.1 | |||
| AK080189.1 | 1 | 2530 | ||
| AK141800.1 | ||||
| AK163006.1 | ||||
| AK220164.1 | BAD90349.1 | |||
| BC062934.1 | AAH62934.1 | |||
| BC064678.1 | AAH64678.1 | |||
| CJ122836.1 | ||||
| CN714704.1 |
More Information
Set Membership: |
|

Genomic Map