Assay Details
Target Gene Details
Entrez Gene ID: | 71586 |
Gene Name: | interferon induced with helicase C domain 1 |
Gene Aliases: |
9130009C22Rik, Helicard, Hlcd, MDA5, RLR-2 |
Location: |
Chr.2:62595612-62646255 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ifih1 | NM_001164477.1 | 8 | 1893 | NP_001157949.1 |
| NM_027835.3 | 9 | 2040 | NP_082111.2 | |
| AF374384.1 | 9 | 1949 | AAM21359.1 | |
| AK037057.1 | 9 | 1938 | BAC29687.2 | |
| AK040519.1 | 9 | 1997 | BAC30614.1 | |
| AK153018.1 | 9 | 1947 | BAE31652.1 | |
| AK153344.1 | BAE31920.1 | |||
| AY075132.1 | 9 | 1715 | AAL77205.1 | |
| BC004031.1 | 2 | 182 | AAH04031.1 | |
| BC025508.1 | 4 | 484 | AAH25508.1 | |
| BC080200.1 | 8 | 1798 | AAH80200.1 | |
| EF113547.1 | 9 | 1729 | ||
| EF113548.1 | 9 | 1729 | ||
| EU084750.1 | 9 | 1729 |

Genomic Map