Assay Details
Target Gene Details
Entrez Gene ID: | 70789 |
Gene Name: | kynureninase (L-kynurenine hydrolase) |
Gene Aliases: |
4432411A05Rik |
Location: |
Chr.2:43555258-43683048 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kynu | NM_001289593.1 | NP_001276522.1 | ||
| NM_001289594.1 | NP_001276523.1 | |||
| NM_027552.2 | NP_081828.1 | |||
| AK014484.1 | BAB29386.2 | |||
| AK040584.1 | BAE20583.1 | |||
| AK050024.1 | BAC34035.1 | |||
| BC026950.1 | AAH26950.1 | |||
| BC069848.1 | AAH69848.1 |
More Information
Set Membership: |
|

Genomic Map