Assay Details
Target Gene Details
Entrez Gene ID: | 22343 |
Gene Name: | lin-7 homolog C (C. elegans) |
Gene Aliases: |
9130007B12Rik, AI303698, AU019331, AW125731, D2Ertd520e, LIN-7-C, LIN-7C, MALS-3, Veli3 |
Location: |
Chr.2:109890878-109900937 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lin7c | NM_011699.3 | NP_035829.1 | ||
| AF087695.1 | AAC78483.1 | |||
| AF173083.1 | AAD48502.1 | |||
| AK015399.1 | BAB29830.1 | |||
| AK078923.1 | BAC37462.1 | |||
| AK083299.1 | ||||
| AK128989.1 | ||||
| AK133531.1 | ||||
| AK135810.1 | BAE22670.1 | |||
| AK161076.1 | BAE36186.1 | |||
| BC046966.1 | AAH46966.1 |
More Information
Set Membership: |
|

Genomic Map