Assay Details
Target Gene Details
Entrez Gene ID: | 14725 |
Gene Name: | low density lipoprotein receptor-related protein 2 |
Gene Aliases: |
AI315343, AW536255, D230004K18Rik, Gp330, Megalin, b2b1625.2Clo |
Location: |
Chr.2:69424335-69586067 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 76 - Exon 77 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lrp2 | NM_001081088.1 | NP_001074557.1 | ||
| AF197160.1 | AAF61488.1 | |||
| AK166702.1 | BAE38957.1 |
More Information
Set Membership: |
|

Genomic Map