Assay Details
Target Gene Details
Entrez Gene ID: | 66634 |
Gene Name: | minichromosome maintenance deficient 8 (S. cerevisiae) |
Gene Aliases: |
5730432L01Rik |
Location: |
Chr.2:132816141-132844197 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mcm8 | NM_001291054.1 | NP_001277983.1 | ||
| NM_025676.4 | NP_079952.2 | |||
| AK012845.1 | BAB28507.1 | |||
| AK017607.1 | ||||
| AK019268.1 | BAB31638.1 | |||
| AK133858.1 | BAE21892.1 | |||
| BC046780.1 | AAH46780.1 | |||
| BC052070.1 | AAH52070.1 | |||
| BY766288.1 |
More Information
Set Membership: |
|

Genomic Map