Assay Details
Target Gene Details
Entrez Gene ID: | 17005 |
Gene Name: | leukocyte tyrosine kinase |
Gene Aliases: |
- |
Location: |
Chr.2:119751326-119760431 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ltk | NM_008523.2 | NP_032549.2 | ||
| NM_203345.2 | NP_976220.2 | |||
| NM_206941.1 | NP_996824.1 | |||
| NM_206942.2 | NP_996825.2 | |||
| AK157478.1 | ||||
| BC047138.1 | ||||
| BC141299.1 | ||||
| M90470.1 | AAA39451.1 | |||
| X07984.1 | CAA30793.1 | |||
| X52621.1 | CAA36848.1 |
More Information
Set Membership: |
|

Genomic Map