Assay Details
Target Gene Details
Entrez Gene ID: | 56846 |
Gene Name: | N-terminal EF-hand calcium binding protein 3 |
Gene Aliases: |
2900010M17Rik, AI853434, Apba2bp, Nip1, XB51 |
Location: |
Chr.2:154544399-154559406 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Necab3 | NM_021546.3 | NP_067521.2 | ||
| AK013520.1 | BAB28895.1 | |||
| AK044296.1 | BAC31859.1 | |||
| AK158225.1 | BAE34411.1 |

Genomic Map