Assay Details
Target Gene Details
Entrez Gene ID: | 17979 |
Gene Name: | nuclear receptor coactivator 3 |
Gene Aliases: |
2010305B15Rik, AW321064, Actr, Aib1, KAT13B, Rac3, Src3, Tram-1, Tram1, bHLHe42, p/Cip, pCip |
Location: |
Chr.2:165992637-166073242 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 9 - Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ncoa3 | NM_008679.3 | NP_032705.2 | ||
| AF000581.1 | AAC05020.1 | |||
| AK137507.1 | BAE23388.1 | |||
| AK142306.1 | BAE25022.1 | |||
| AK156345.1 | ||||
| BC086482.1 | ||||
| BC093478.1 | ||||
| BC141177.1 |
More Information
Set Membership: |
|

Genomic Map