Assay Details
Target Gene Details
Entrez Gene ID: | 68375 |
Gene Name: | NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 8 |
Gene Aliases: |
0610033L03Rik, AW261656 |
Location: |
Chr.2:36036334-36049292 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ndufa8 | NM_026703.2 | 1 | 57 | NP_080979.1 |
| AK002744.1 | 1 | 57 | BAB22322.1 | |
| AK088463.1 | 1 | 105 | ||
| BC012416.1 | 1 | 55 | AAH12416.1 | |
| BC024673.1 | 1 | 54 | AAH24673.1 |
More Information
Set Membership: |
|

Genomic Map