Assay Details
Target Gene Details
Entrez Gene ID: | 228829 |
Gene Name: | PHD finger protein 20 |
Gene Aliases: |
6820402O20Rik |
Location: |
Chr.2:156196647-156309953 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Phf20 | NM_172674.2 | NP_766262.2 | ||
| AK033017.1 | BAC28129.1 | |||
| AK038573.1 | BAC30050.2 | |||
| AK045309.1 | BAC32304.1 | |||
| BC060121.1 | AAH60121.1 |

Genomic Map