Assay Details
Target Gene Details
Entrez Gene ID: | 78928 |
Gene Name: | phosphatidylinositol glycan anchor biosynthesis, class T |
Gene Aliases: |
2510012P17Rik, 4930534E15Rik, CGI-06, NDAP, Ndap7 |
Location: |
Chr.2:164497511-164508301 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 9 - Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pigt | NM_133779.2 | NP_598540.2 | ||
| AK019717.2 | ||||
| AK044499.1 | BAC31951.1 | |||
| AK049171.1 | ||||
| AK049990.1 | ||||
| AK090020.1 | 9 | 1253 | ||
| AK153462.1 | BAE32014.1 | |||
| AK153464.1 | BAE32015.1 | |||
| AK155057.1 | BAE33018.1 | |||
| AK157226.1 | BAE34008.1 | |||
| AK162573.1 | 7 | 1459 | ||
| AK177432.1 | ||||
| BC034175.1 | AAH34175.1 |

Genomic Map