Assay Details
Target Gene Details
Entrez Gene ID: | 14827 |
Gene Name: | protein disulfide isomerase associated 3 |
Gene Aliases: |
58kDa, ERp57, ERp60, ERp61, Erp, Grp58, PDI, PDI-Q2, PI-PLC, PLC[a], Plca |
Location: |
Chr.2:121413902-121438687 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pdia3 | NM_007952.2 | NP_031978.2 | ||
| AK088721.1 | BAC40527.1 | |||
| AK132459.1 | ||||
| AK133799.1 | BAE21849.1 | |||
| AK167807.1 | BAE39834.1 | |||
| AK169611.1 | BAE41259.1 | |||
| BC003285.1 | AAH03285.1 | |||
| BC033439.1 | AAH33439.1 | |||
| M73329.1 | AAA39944.1 |
More Information
Set Membership: |
|

Genomic Map