Assay Details
Target Gene Details
Entrez Gene ID: | 67005 |
Gene Name: | polymerase (RNA) III (DNA directed) polypeptide K |
Gene Aliases: |
12.3kDa, 1500004O14Rik, AI196849, AU014893, C11, RPC10, RPC11 |
Location: |
Chr.2:181864360-181870830 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Polr3k | NM_025901.3 | NP_080177.1 | ||
| AK005152.1 | BAB23846.1 | |||
| AK008637.1 | ||||
| AK010875.1 | BAB27239.1 | |||
| AK021353.1 | BAB32384.1 | |||
| AK031110.1 | BAC27257.1 | |||
| AK088206.1 | BAC40210.2 | |||
| BY003788.1 |
More Information
Set Membership: |
|

Genomic Map