Assay Details
Target Gene Details
Entrez Gene ID: | 19246 |
Gene Name: | protein tyrosine phosphatase, non-receptor type 1 |
Gene Aliases: |
PTP-1B, PTP-HA2, PTP1B |
Location: |
Chr.2:167932303-167979385 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ptpn1 | NM_011201.3 | 1 | 113 | NP_035331.3 |
| AK149979.1 | 1 | 69 | BAE29209.1 | |
| AK150322.1 | 1 | 69 | BAE29467.1 | |
| AK151371.1 | ||||
| AK153088.1 | 1 | 52 | ||
| AK156281.1 | 1 | 73 | ||
| AK157431.1 | 1 | 114 | BAE34088.1 | |
| AK165567.1 | 1 | 78 | ||
| AK165949.1 | 1 | 69 | ||
| AK166020.1 | 1 | 69 | BAE38525.1 | |
| AK168255.1 | 1 | 51 | ||
| AK168772.1 | 1 | 68 | BAE40608.1 | |
| AK170620.1 | 1 | 69 | BAE41915.1 | |
| AK171380.1 | 1 | 69 | BAE42421.1 | |
| AK172206.1 | 1 | 62 | BAE42881.1 | |
| BC010191.1 | 1 | 31 | AAH10191.1 | |
| M97590.1 | 1 | 29 |
More Information
Set Membership: |
|

Genomic Map