Assay Details
Target Gene Details
Entrez Gene ID: | 19246 |
Gene Name: | protein tyrosine phosphatase, non-receptor type 1 |
Gene Aliases: |
PTP-1B, PTP-HA2, PTP1B |
Location: |
Chr.2:167932303-167979385 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ptpn1 | NM_011201.3 | NP_035331.3 | ||
| AK149979.1 | BAE29209.1 | |||
| AK150322.1 | BAE29467.1 | |||
| AK151371.1 | ||||
| AK153088.1 | ||||
| AK156281.1 | ||||
| AK157431.1 | BAE34088.1 | |||
| AK165567.1 | ||||
| AK165949.1 | ||||
| AK166020.1 | BAE38525.1 | |||
| AK168255.1 | ||||
| AK168772.1 | BAE40608.1 | |||
| AK170620.1 | BAE41915.1 | |||
| AK171380.1 | BAE42421.1 | |||
| AK172206.1 | BAE42881.1 | |||
| AK185938.1 | ||||
| BC010191.1 | AAH10191.1 | |||
| M97590.1 |
More Information
Set Membership: |
|

Genomic Map