Assay Details
Target Gene Details
Entrez Gene ID: | 94249 |
Gene Name: | solute carrier family 24 (sodium/potassium/calcium exchanger), member 3 |
Gene Aliases: |
NCKX3 |
Location: |
Chr.2:145167712-145642166 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc24a3 | NM_053195.2 | NP_444425.1 | ||
| AF314821.1 | AAG60049.1 | |||
| BC017615.1 | AAH17615.1 |
More Information
Set Membership: |
|

Genomic Map