Assay Details
Target Gene Details
Entrez Gene ID: | 56324 |
Gene Name: | signal transducing adaptor molecule (SH3 domain and ITAM motif) 2 |
Gene Aliases: |
1200004O12Rik, 5730456G07Rik, BB145015, Hbp |
Location: |
Chr.2:52692198-52742149 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Stam2 | NM_019667.2 | 7 | 657 | NP_062641.1 |
| AB012611.1 | 7 | 672 | BAA33547.1 | |
| AK004604.2 | 7 | 657 | BAB23403.1 | |
| AK044230.1 | 6 | 586 | BAC31830.1 | |
| AK078244.1 | 7 | 689 | ||
| AK159699.1 | 7 | 686 | BAE35299.1 | |
| AK168729.1 | 7 | 689 | BAE40570.1 | |
| BC013818.1 | 7 | 693 | AAH13818.1 | |
| DQ141211.1 | 8 | 887 | AAZ82831.1 |
More Information
Set Membership: |
|

Genomic Map