Assay Details
Target Gene Details
Entrez Gene ID: | 329384 |
Gene Name: | peptidyl-tRNA hydrolase 1 homolog (S. cerevisiae) |
Gene Aliases: |
2210013M04Rik, AW046744 |
Location: |
Chr.2:32775738-32777593 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ptrh1 | NM_178595.3 | NP_848710.1 | ||
| AK075833.1 | BAC35994.1 | |||
| BC098218.1 | AAH98218.1 |
More Information
Set Membership: |
|

Genomic Map