Assay Details
Target Gene Details
Entrez Gene ID: | 78830 |
Gene Name: | solute carrier family 25 (mitochondrial carrier, Aralar), member 12 |
Gene Aliases: |
2610002D09Rik, AI839531, B230107K20Rik, BB129864 |
Location: |
Chr.2:71274270-71376694 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc25a12 | NM_172436.3 | NP_766024.1 | ||
| AK019150.1 | ||||
| AK030595.1 | BAC27037.1 | |||
| AK083156.1 | BAC38787.1 | |||
| AK144958.1 | ||||
| AK147948.1 | ||||
| BC060505.1 | AAH60505.1 |
More Information
Set Membership: |
|

Genomic Map