Assay Details
Target Gene Details
Entrez Gene ID: | 227731 |
Gene Name: | solute carrier family 25 (mitochondrial carrier, phosphate carrier), member 25 |
Gene Aliases: |
1110030N17Rik, MCSC, mKIAA1896 |
Location: |
Chr.2:32414487-32451470 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc25a25 | NM_001164357.1 | NP_001157829.1 | ||
| NM_001164358.1 | NP_001157830.1 | |||
| NM_001290558.1 | NP_001277487.1 | |||
| NM_146118.3 | NP_666230.2 | |||
| AK049392.1 | BAC33730.1 | |||
| AK082756.1 | BAC38604.1 | |||
| AK122568.2 | BAC65850.1 | |||
| AK132204.1 | BAE21031.1 | |||
| BC019978.1 | AAH19978.1 | |||
| BC022114.1 | AAH22114.1 | |||
| BC037109.1 | AAH37109.1 | |||
| BC066998.1 | AAH66998.1 |
More Information
Set Membership: |
|

Genomic Map