Assay Details
Target Gene Details
Entrez Gene ID: | 227717 |
Gene Name: | pyroglutamylated RFamide peptide |
Gene Aliases: |
P518 |
Location: |
Chr.2:31806168-31810518 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Qrfp | NM_183424.3 | NP_906269.1 | ||
| AK029144.1 | BAC26323.1 |

Genomic Map