Assay Details
Target Gene Details
Entrez Gene ID: | 57138 |
Gene Name: | solute carrier family 12, member 5 |
Gene Aliases: |
KCC2, mKIAA1176 |
Location: |
Chr.2:164960818-164999731 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc12a5 | NM_020333.2 | 17 | 2508 | NP_065066.2 |
| AF332063.1 | 17 | 2178 | AAK56092.1 | |
| AF332064.1 | 17 | 2178 | AAK56093.1 | |
| AK122460.1 | 17 | 2242 | BAC65742.1 | |
| AK147262.1 | 17 | 2510 | BAE27805.1 | |
| AK165769.1 | BAE38372.1 | |||
| BC054808.1 | 17 | 2174 | AAH54808.1 | |
| KJ535320.1 | 17 | 2199 | ||
| KJ535321.1 | 17 | 2199 | ||
| KJ535322.1 | 17 | 2355 |
More Information
Set Membership: |
|

Genomic Map