Assay Details
Target Gene Details
Entrez Gene ID: | 170791 |
Gene Name: | RNA binding motif protein 39 |
Gene Aliases: |
1500012C14Rik, 2310040E03Rik, B330012G18Rik, C79248, R75070, Rnpc2, caper |
Location: |
Chr.2:156147239-156180238 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rbm39 | NM_001291114.1 | NP_001278043.1 | ||
| NM_001291115.1 | NP_001278044.1 | |||
| NM_133242.3 | NP_573505.2 | |||
| AK134091.1 | BAE22009.1 | |||
| AK135289.1 | BAE22477.1 | |||
| AK146046.1 | ||||
| AK147076.1 | BAE27657.1 | |||
| AK154989.1 | BAE32977.1 | |||
| BC004000.1 | AAH04000.1 | |||
| BC030493.1 | AAH30493.1 | |||
| BC082607.1 | ||||
| BC086645.1 | AAH86645.1 |
More Information
Set Membership: |
|

Genomic Map