Assay Details
Target Gene Details
Entrez Gene ID: | 12258 |
Gene Name: | serine (or cysteine) peptidase inhibitor, clade G, member 1 |
Gene Aliases: |
C1INH, C1nh |
Location: |
Chr.2:84765360-84775445 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Serping1 | NM_009776.3 | 5 | 895 | NP_033906.2 |
| AF010254.1 | 5 | 870 | AAC40136.1 | |
| AK144292.1 | 4 | 856 | ||
| AK165744.1 | 4 | 843 | ||
| AK167585.1 | 4 | 832 | ||
| BC002026.1 | 4 | 826 | AAH02026.1 | |
| Y10386.1 | 5 | 852 | CAA71412.1 |
More Information
Set Membership: |
|

Genomic Map